Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfamethazine, 99%, Thermo Scientific Chemicals

Catalog No. p-194284
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-68-1
Molecular Formula C12H14N4O2S
Molecular Weight (g/mol) 278.33
MDL Number MFCD00006066
InChI Key ASWVTGNCAZCNNR-UHFFFAOYSA-N
Synonym sulfamethazine, sulfadimidine, sulfadimethyldiazine, sulfamezathine, sulfadimerazine, sulphamethazine, sulphamezathine, sulfadimezine, sulphadimidine, sulfadimesin
PubChem CID 5327
ChEBI CHEBI:102265
IUPAC Name 4-amino-N-(4,6-dimethylpyrimidin-2-yl)benzenesulfonamide
SMILES CC1=CC(=NC(=N1)NS(=O)(=O)C2=CC=C(C=C2)N)C

Specifications

Melting Point 197.0°C to 200.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 9047
Formula Weight 278.32
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Sulfamethazine, 99%
Show More Show Less
Hazard Category LIGHT SENSITIVE
EINECSNumber 200-346-4
RTECSNumber WO9275000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.