Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Sulfamethazine Recombinant Rabbit Monoclonal Antibody (HuaM009-51B)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA544677
This item is not returnable.
View return policy
Description
Application notes: Indirect ELISA.

Specifications
Sulfamethazine | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide; pH 7.4 | |
Sulfadimidine. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Chemical |
ELISA | |
HuaM009-51B | |
Unconjugated | |
Rabbit | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction