Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfamic Acid, ACS, 99.3-100.3%, Spectrum™ Chemical
SDP

Catalog No. 18614152
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle

Chemical Identifiers

CAS 5329-14-6
Molecular Formula H3NO3S
Molecular Weight (g/mol) 97.09
InChI Key IIACRCGMVDHOTQ-UHFFFAOYSA-N
IUPAC Name sulfamic acid
SMILES NS(O)(=O)=O

Specifications

CAS Max % 100%
Heavy Metals 0.001%
Packaging Poly Bottle
Residue after Ignition 0.01%
Quantity 500 g
Percent Purity 99.3 to 100.3%
Grade ACS

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.