Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfamic Acid, ACS Reagent, Reagents

Catalog No. 01337574
Click to view available options
Quantity:
100 mL
500 g
2.5 kg
6 lb.
10 kg
50 lb.

Sulfamic Acid, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 5329-14-6
Molecular Formula H3NO3S
Molecular Weight (g/mol) 97.09
InChI Key IIACRCGMVDHOTQ-UHFFFAOYSA-N
Synonym Amidosulfonic Acid, Aminosulfonic Acid, Sulphamic Acid
IUPAC Name sulfamic acid
SMILES NS(O)(=O)=O

Specifications

Color White to Off-White
Melting Point 205°C
Packaging Poly Pail
Specific Gravity 2.15
Quantity 10 kg
Physical Form Solid
Grade ACS
Chemical Name or Material Sulfamic Acid, ACS Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.