Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfamide, 98+%

Catalog No. AC199021000
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 7803-58-9 | H4N2O2S | 96.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7803-58-9
Molecular Formula H4N2O2S
Molecular Weight (g/mol) 96.10
MDL Number MFCD00011606
InChI Key NVBFHJWHLNUMCV-UHFFFAOYSA-N
Synonym sulfuric diamide, sulfamamide, sulfuryl amide, sulfonyl diamide, sulfuryl diamide, sulfamoylamine, sulphamide, sulphuric diamide, unii-vs7tzw634v, imidosulfamic acid
PubChem CID 82267
ChEBI CHEBI:29368
IUPAC Name sulfamoylamine
SMILES NS(N)(=O)=O

Specifications

Melting Point 87.0°C to 91.0°C
Color White
Density 1.6100g/mL
pH 5.0 to 7.0 (10% soln. aq. soln.)
Elemental Analysis 98.0% min.
Loss on Drying 0.5% (50°C, 2 hrs) (vacuum) max.
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass Bottle
Linear Formula H2NSO2NH2
Quantity 100 g
Specific Gravity 1.61
Merck Index 15,9054
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone,hot alcohol
Formula Weight 96.11
Percent Purity 98+%
Related Substances (TLC) Passes test
Physical Form Crystalline Powder
Chemical Name or Material Sulfamide
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 232-262-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.