Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) acetate trihydrate, 99+%, for analysis

Catalog No. AC201755000
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle

CAS: 6080-56-4 | C4H12O7Pb | 379.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6080-56-4
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
PubChem CID 22456
ChEBI CHEBI:33112
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 75.0°C
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 500 g
Assay Percent Range 99.0 to 103% (Complexometry)
Linear Formula (CH3CO2)2Pb·3H2O
Merck Index 15, 5452
Solubility Information Solubility in water: 625g/L. Other solubilities: 1g/30mL alcohol, freely soluble in glycerol
IUPAC Name lead(2+);diacetate;trihydrate
Formula Weight 379.33
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Lead(II) acetate trihydrate, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause damage to organs through prolonged or repeated exposure if swallowed.
May damage the unborn child.
Suspected of damaging fertility.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHA
RTECSNumber OF8050000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.