Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfanilamide Reagent, 1% (w/v) in 10% (v/v) Hydrochloric Acid, Ricca Chemical
SDP

Catalog No. 810316 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle

Chemical Identifiers

CAS 63-74-1, 7647-01-0, 7732-18-5
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.202
InChI Key FDDDEECHVMSUSB-UHFFFAOYSA-N
Synonym sulfanilamide, sulphanilamide, sulfamine, sulphonamide, p-aminobenzenesulfonamide, sulfonylamide, sulfonamide, p-aminobenzenesulfamide, bacteramid, streptasol
PubChem CID 5333
ChEBI CHEBI:45373
IUPAC Name 4-aminobenzenesulfonamide
SMILES C1=CC(=CC=C1N)S(=O)(=O)N

Specifications

Color Colorless
Percent Purity 10%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix 10% (v/v) Hydrochloric Acid
pH <1
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 1% (w/v)
Chemical Name or Material Sulfanilamide Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.