Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Sulfatase Modifying Factor 2/ SUMF2 Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. AF2779 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2779 100 μg
AF2779SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2779 Supplier R&D Systems Supplier No. AF2779
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody

Sulfatase Modifying Factor 2/SUMF2 Polyclonal specifically detects Sulfatase Modifying Factor 2/SUMF2 in Human samples. It is validated for Immunoprecipitation∣Western Blot.

Specifications

Antigen Sulfatase Modifying Factor 2/SUMF2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Gene Accession No. Q8NBJ7
Gene Alias C-alpha-formyglycine-generating enzyme 2, C-alpha-formylglycine-generating enzyme 2, DKFZp566I1024, DKFZp686I1024, DKFZp686L17160, DKFZp781L1035, FGE2, MGC99485, paralog of the formylglycine-generating enzyme, pFGE, sulfatase modifying factor 2, sulfatase-modifying factor 2
Gene Symbols SUMF2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Sulfatase Modifying Factor 2/SUMF2 Gln26-Leu301 Accession # Q8NBJ7
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25870
Test Specificity Detects human Sulfatase Modifying Factor 2/SUMF2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 45% cross-reactivity with recombinant mouse SUMF2 is observed and less than 1% cross-reactivity with recombinant human SUMF1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Type Antibody
Isotype IgG
Product Line R&D Systems
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.