Learn More
Thermo Scientific™ Sulfo-SBED Biotin Label Transfer Reagent, 10 mg PROMO
Description
Ready-to-use kit eliminates the need to prepare the necessary buffers or purchase the critical reagents necessary to perform a label transfer experiment. Key reagents, Sulfo-SBED and DTT, are supplied in unique, no-weigh, single-use packaging.
Highlights:
- Sulfo-NHS ester reacts with primary amines at pH 7-9
- Aryl azide conjugates randomly when activated with UV light
- Cleavable disulfide bond allows biotin label transfer
- Discover protein interactions without radiolabeling
Includes:
No-Weigh Sulfo-SBED Label Transfer Reagent, 8 x 1mg; BupH Phosphate Buffered Saline, 1 Pk. (makes 500 mL); Label Transfer Buffer (20X), 200 mL (makes 4L); Streptavidin-HRP Conjugate, 0.1mg; No-Weigh Dithiothreitol (DTT), 8 x 7.7mg; Slide-A-Lyzer MINI Dialysis Units plus Float, 10K MWCO, 10-100μL, 10 units/Pk.
Specifications
Specifications
Format | Powder |
Quantity | 10 mg |
Product Type | Crosslinker |
Solubility | Water |
Content And Storage | Upon receipt store at 4°C. |
Labeling Method | Chemical Labeling, Label Transfer |
Reactive Moiety | Sulfo-NHS Ester, Aryl Azide |
Product Line | Pierce™ |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.