Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfonfluorescein 75.0+%, TCI America™
SDP

Catalog No. p-7135828 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S01321G 1 g
S01325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S01321G Supplier TCI America Supplier No. S01321G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4424-03-7
Molecular Formula C19H12O6S
Molecular Weight (g/mol) 368.36
MDL Number MFCD00070618
InChI Key VJYGFSGAAOVTLC-UHFFFAOYSA-N
PubChem CID 78140
IUPAC Name 1,1-dioxospiro[2,1$l^{6}-benzoxathiole-3,9'-xanthene]-3',6'-diol
SMILES C1=CC=C2C(=C1)C3(C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O)OS2(=O)=O

Specifications

Quantity 1 g
Formula Weight 368.36
Percent Purity ≥75.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sulfonfluorescein
RTECSNumber WG9930000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.