Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulforhodamine 101 acid chloride

Catalog No. aaj60581ma
Click to view available options
Quantity:
10 mg

CAS: 82354-19-6 | C31H29ClN2O6S2 | 625.15 g/mol

Fluorescent labeling compound

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82354-19-6
Molecular Formula C31H29ClN2O6S2
Molecular Weight (g/mol) 625.15
MDL Number MFCD00012406
InChI Key MPLHNVLQVRSVEE-UHFFFAOYSA-N
Synonym Texas Red™; Sulforhodamine 101 sulfonyl chloride
PubChem CID 452705
ChEBI CHEBI:51247
IUPAC Name 16-[4-(chlorosulfonyl)-2-sulfonatophenyl]-3-oxa-9λ⁵,23-diazaheptacyclo[17.7.1.1⁵,⁹.0²,¹⁷.0⁴,¹⁵.0²³,²⁷.0¹³,²⁸]octacosa-1,4,9(28),13,15,17,19(27)-heptaen-9-ylium
SMILES [O-]S(=O)(=O)C1=C(C=CC(=C1)S(Cl)(=O)=O)C1=C2C=C3CCC[N+]4=C3C(CCC4)=C2OC2=C3CCCN4CCCC(C=C12)=C34

Specifications

Beilstein 8667894
Formula Weight 625.15
Sensitivity Light and moisture sensitive
Color Red
Quantity 10 mg
Physical Form Powder
Chemical Name or Material Sulforhodamine 101 acid chloride
Hazard Category H301-H332-H351
Hazard Statement GHS H Statement
H351-H373-H302
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P330-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.