Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulforhodamine B, MP Biomedicals™

Catalog No. ICN15247905 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15247905 5 g
ICN15247901 1 g
ICN15247925 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15247905 Supplier MP Biomedicals, Inc Supplier No. 0215247905
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3520-42-1
Molecular Formula C29H33N2NaO7S2
Molecular Weight (g/mol) 608.70
MDL Number MFCD00010180
InChI Key XJENLUNLXRJLEZ-UHFFFAOYSA-M
Synonym Acid Red 52, Phloxinrhodamine
PubChem CID 131852807
IUPAC Name sodium 3,6-bis(diethylamino)-9-(2,4-disulfonatophenyl)-2,7-dimethyl-10λ⁴-xanthen-10-ylium
SMILES [Na+].CCN(CC)C1=CC2=[O+]C3=CC(N(CC)CC)=C(C)C=C3C(C3=CC=C(C=C3S([O-])(=O)=O)S([O-])(=O)=O)=C2C=C1C

Specifications

Color Brown-Purple
Quantity 5 g
Formula Weight 580.66
Physical Form Powder
Chemical Name or Material Sulforhodamine B
EINECSNumber 222-529-8
Recommended Storage Store at room temperature.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.