Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sulfosalicylic Acid, 10% w/v, Reagents
Click to view available options
Quantity:
500 mL
1 L
4 L
Specifications
Specifications
Color | Colorless to Light Pink |
pH | <1 |
Boiling Point | 101°C |
Packaging | Glass Bottle |
Quantity | 4 L |
Specific Gravity | 1.0446 |
Percent Purity | 10% w/v |
Grade | Reagent |
Physical Form | Liquid |
Chemical Name or Material | Sulfosalicylic Acid, 10% w/v |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction