Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfosalicylic Acid, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2kg
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

5965-83-3
C7H10O8S
254.209
BHDKTFQBRFWJKR-UHFFFAOYSA-N
2723734

High quality chemicals for laboratory and specialized industrial use

Quantity 500g
SMILES C1=CC(=C(C=C1S(=O)(=O)O)C(=O)O)O.O.O
IUPAC Name 2-hydroxy-5-sulfobenzoic acid;dihydrate
Packaging Wide Mouth Amber Glass Bottle
Formula Weight 254.22
Physical Form Crystals
Chemical Name or Material Sulfosalicylic Acid, Dihydrate
Grade Baker Analyzed™ ACS Reagent