Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sulfotyrosine Antibody (7C5), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$646.00
Specifications
Antigen | Sulfotyrosine |
---|---|
Clone | 7C5 |
Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 |
Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
Sulfotyrosine Monoclonal specifically detects Sulfotyrosine in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Sulfotyrosine | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 | |
Monoclonal | |
Purified | |
RUO | |
IgG1 | |
Protein G purified |
7C5 | |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Synthetic Sulfotyrosine conjugated to Keyhole Limpet Hemocyanin (KLH). | |
Primary | |
Specific for sulfated tyrosine residues on various proteins. Does not detect free Sulfotyrosine. Does not cross-react with phosphotyrosine. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title