Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Sulfotyrosine Monoclonal Antibody (7C5), PerCP
GREENER_CHOICE

Catalog No. PIMA545835
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545835 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545835 Supplier Invitrogen™ Supplier No. MA545835
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

A 1:1,000 dilution of MA5-45835 was sufficient for detection of Sulfotyrosine in 0.5 μg of Sulfotyrosine conjugated to BSA by ECL immunoblot analysis using Goat Anti-Mouse IgG:HRP as the secondary Antibody. Specific for sulfated tyrosine residues on various proteins. Does not detect free Sulfotyrosine. Does not cross-react with phosphotyrosine.

Sulfotyrosine is involved in protein-protein interactions. Sulfotyrosines are present in all multicellular eukaryotes, but unicellular eukaryotes such as yeast and prokaryotes are not capable of tyrosine sulfation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sulfotyrosine
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 7C5
Concentration 1 mg/mL
Conjugate PerCP
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene Alias sTYR; S-Tyrosine; Sulfated Tyrosine; Sulfotyrosine; Sulphated Tyrosine; Sulphotyrosine; Tyrosine sulfation; Tyrosine Sulphation
Host Species Mouse
Immunogen Synthetic Sulfotyrosine conjugated to Keyhole Limpet Hemocyanin (KLH).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.