Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfotyrosine Mouse anti-Human, ATTO 390, Clone: 7C5, Abnova™

Catalog No. 89338857 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-338-857 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-338-857 Supplier Abnova Corporation Supplier No. MAB19353
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

Sulfotyrosine Monoclonal antibody specifically detects Sulfotyrosine in Human samples. It is validated for WB, ELISA, ICC, IF.

Specifications

Antigen Sulfotyrosine
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 7C5
Conjugate Atto 390
Host Species Mouse
Immunogen Synthetic Sulfotyrosine conjugated to Keyhole Limpet Hemocyanin (KLH)
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20°C
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.