Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfur, 99.999%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
7704-34-9
S
32.05
MFCD00085316
NINIDFKCEFEMDL-UHFFFAOYSA-N
polymeric,lac,ad usum externum,acmc-20a47y,rubbervulcanizationagentis ot
5362487
CHEBI:26833
sulfur
[S]
7704-34-9
S
32.05
MFCD00085316
NINIDFKCEFEMDL-UHFFFAOYSA-N
polymeric,lac,ad usum externum,acmc-20a47y,rubbervulcanizationagentis ot
5362487
CHEBI:26833
sulfur
[S]
For ultra trace analysis
Assay Percent Range 99.999%
Fieser 01,1118; 03,273; 05,632; 06,556; 14,291; 15,297
Merck Index 15, 9099
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in alcohol and ether,soluble in carbon disulfide,benzene,(about 2.4%,at 30°C,much more at higher temps.),toluene,,acetone (2.65% at 25°C),chloroform (1.5% at 18°C)
Formula Weight 32.05
Physical Form Pieces (12mm Pieces and Smaller)
Percent Purity 99.999%
Grade Trace Metal Basis
Density 2.36
Packaging Glass bottle
Solubility Solubility in water: insoluble. Other solubilities: slightly soluble in alcohol and ether, soluble in carbon disulfide, benzene, (about 2.4%, at 30°C, much more at higher temps.),toluene,, acetone (2.65% at 25°C), chloroform (1.5% at 18°C)
Total Trace Metal Impurities 0.001% max
Boiling Point 445.0°C
Color Yellow
Melting Point 114.0°C
Quantity 50g
Flash Point 168°C
Chemical Name or Material Sulfur, 99.999%
Show More Show Less

GHS H Statement
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and w

GHS Signal Word: Warning

EINECSNumber : 231-722-6

RTECSNumber : WS4250000

TSCA : TSCA