Learn More
Eisco™ Sulfur Mineral Specimen
SCI_ED
Catalog No. S24712 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S24712 1 Ea. 2 to 3 cm
S26600 10/Pk. 2 to 3 cm
S24713 12/Pk. 2 to 3 cm
S26647 1 Ea. 7 to 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S24712 Supplier Eisco™ Supplier No. ESNG0026
Add to Cart
Add to Cart
Sulfur Mineral Specimen ideal for mineral identification exercises, due to its instantly recognizable physical properties.
  • For use in learning about streak tests
  • Sulfur leaves an unexpected white streak and distinct smell
  • For use in classroom identification labs and exercises
  • Correlates well to Next Generation Science Standards Evidence Statements
Specifications
Product Type Mineral
For Use With (Application) Lessons on identifying minerals in unconventional ways
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm
Specimen Type Sulfur