Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sulfuric Acid, 0.02N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
1 L
4 L
Packaging:
Cubetainer
Narrow Mouth Polyethylene Bottle
Specifications
Specifications
Packaging | Narrow Mouth Polyethylene Bottle |
Specific Gravity | 1L = 1.02kg |
Quantity | 1 L |
Formula Weight | 98.08 |
Grade | BAKER ANALYZED™ Reagent |
Chemical Name or Material | Sulfuric Acid, 0.02N Volumetric Solution |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction