Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 0.0500 N (N/20), Ricca Tightrant™
SDP

Catalog No. 821932 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
8219-32 1 L Natural Poly Bottle
R8219000500 500 mL Natural Poly Bottle
R82190004F 4 L Cubitainer
8219-1 4 L Natural Poly Bottle
8219-5 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 8219-32 Supplier Ricca Chemical Company Supplier No. 821932
Add to Cart
Add to Cart
  • Tested in and ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9

Specifications

Packaging Natural Poly Bottle
Quantity 1 L
Synonym dihydrogen oxide, dihydrogen monoxide
Molecular Weight (g/mol) 98.07 g/mol
Grade Laboratory
Chemical Name or Material SULFURIC ACID
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.