Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 0.25 N, AquaPhoenix Scientific™
SCI_ED

Catalog No. S88735 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle,Yellow Cap
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S88735 500 mL Poly Bottle,Yellow Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S88735 Supplier Aqua Phoenix Scientific Supplier No. SA7575P
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7664-93-9 | H2O4S | 98.07 g/mol

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Concentration 0.25 N
Packaging Poly Bottle,Yellow Cap
Quantity 500 mL
Chemical Name or Material Sulfuric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.