Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sulfuric Acid, 0.417 Normal, Reagents
Click to view available options
Quantity:
10 L
20 L
Specifications
Specifications
| pH | <1 |
| Color | Colorless |
| Boiling Point | 100°C |
| Packaging | Cubitainer |
| Specific Gravity | 1.0136 |
| Quantity | 20 L |
| Physical Form | Liquid |
| Grade | Reagent |
| Chemical Name or Material | Sulfuric Acid, 0.417 Normal |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction