Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 0.7% (v/v), Ricca Chemical
SDP

Catalog No. 814616 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
8146-16 500 mL Natural Poly Bottle
8146-32 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 8146-16 Supplier Ricca Chemical Company Supplier No. 814616
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Colorless
Concentration 0.7% (v/v)
CAS Min % 97.0, 1.25
CAS Max % 100.0, 1.3
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 500 mL
Physical Form Liquid
Percent Purity 0.7% V/V
Grade Laboratory
Chemical Name or Material Sulfuric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.