Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 1.00 N, Ricca Tightrant™
SDP

Catalog No. 83005 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
10 L
20 L
55 gal.
Packaging:
Cubitainer
Natural Poly Bottle
Poly Drum
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
8300-5 20 L Cubitainer
8300-4 120 mL Natural Poly Bottle
830016 500 mL Natural Poly Bottle
8300-32 1 L Natural Poly Bottle
R83000004F 4 L Cubitainer
8300-1 4 L Natural Poly Bottle
8300-25 10 L Cubitainer
R830000055E 55 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 missing translation for 'options'
Catalog No. 8300-5 missing translation for 'mfr' Ricca Chemical Company missing translation for 'supplierNo' 83005
Add to Cart
Add to Cart
  • Tested in and ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Concentration 1.00N
CAS Min % 93.0, 4.78
CAS Max % 97.0, 4.97
Packaging Cubitainer
Quantity 20 L
Grade Laboratory
Chemical Name or Material Sulfuric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.