Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sulfuric Acid, 10% (v/v), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
10 L
20 L
16 gal.
Packaging:
Cubitainer
Natural Poly Bottle
Poly Drum
Safety-coated Glass Bottle
Specifications
Specifications
| CAS Min % | 89.0, 9.0 |
| CAS Max % | 91.0, 11.0 |
| Packaging | Poly Drum |
| Solvent or Matrix | Aqueous solution |
| Quantity | 16 gal. |
| Percent Purity | 10% V/V |
| Grade | Laboratory |
| Concentration or Composition (by Analyte or Components) | 10%(v/v) |
| Chemical Name or Material | Sulfuric Acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction