Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 10% (v/v), Ricca Chemical
SDP

Catalog No. 13622618 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
10 L
20 L
16 gal.
Packaging:
Cubitainer
Natural Poly Bottle
Poly Drum
Safety-coated Glass Bottle

Chemical Identifiers

CAS 7732-18-5, 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

CAS Min % 89.0, 9.0
CAS Max % 91.0, 11.0
Packaging Cubitainer
Solvent or Matrix Aqueous solution
Quantity 20 L
Percent Purity 10% V/V
Grade Laboratory
Concentration or Composition (by Analyte or Components) 10%(v/v)
Chemical Name or Material Sulfuric Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.