Learn More
Sulfuric acid, 15% fuming, 12-17% free SO{3}, Thermo Scientific Chemicals
Description
Sulfuric acid is a strong dehydrating agent and is applied in many applications like domestic acidic drain cleaners, electrolytes in lead-acid batteries and various cleansing agents. It is useful for mineral processing, fertilizer manufacture, petroleum purification, wastewater processing and chemical synthesis. The catalytic decomposition of sulfuric acid on mixed Cr/Fe oxide shows a useful application in sulfur-iodine cycle for hydrogen production.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 1.9 |
Sensitivity | Hygroscopic |
Assay Percent Range | 15% fuming, 12-17% free SO3 |
Quantity | 500 g |
Physical Form | Liquid |
Solubility Information | Miscible with water. |
Chemical Name or Material | Sulfuric acid; 15% fuming |
Safety and Handling
Hazard Category | H290-H314-H335 |
Precautionary Statement | P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: II |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.