Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric acid, 15% fuming, 12-17% free SO{3}

Catalog No. p-4408637
Click to view available options
Quantity:
500 g

CAS: 8014-95-7 | H2O7S2 | 178.13 g/mol

Sulfuric acid is a strong dehydrating agent and is applied in many applications like domestic acidic drain cleaners, electrolytes in lead-acid batteries and various cleansing agents. It is useful for mineral processing, fertilizer manufacture, petroleum purification, wastewater processing and chemical synthesis. The catalytic decomposition of sulfuric acid on mixed Cr/Fe oxide shows a useful application in sulfur-iodine cycle for hydrogen production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8014-95-7
Molecular Formula H2O7S2
Molecular Weight (g/mol) 178.13
MDL Number MFCD00137792
InChI Key HIFJUMGIHIZEPX-UHFFFAOYSA-N
Synonym oleum, fuming sulfuric acid, sulfuric acid, fuming, oleum iodisum, sulfuric acid fuming, oleum fuming sulfuric acid, sulfuric acid, mixt with sulfur trioxide, sulfur trioxide, mixt. with sulfuric acid, sulfuric acid mixture with sulfur trioxide
PubChem CID 24681
IUPAC Name sulfonylideneoxidane; sulfuric acid
SMILES O=S(=O)=O.OS(O)(=O)=O

Specifications

Density 1.9
Sensitivity Hygroscopic
Assay Percent Range 15% fuming, 12-17% free SO3
Quantity 500 g
Physical Form Liquid
Solubility Information Miscible with water.
Chemical Name or Material Sulfuric acid; 15% fuming
Hazard Category H290-H314-H335
Precautionary Statement P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.