Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 18.0 N, Ricca Chemical
SDP

Catalog No. 8343185PT Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Packaging:
Natural Poly Bottle
Safety-coated Glass Bottle

Chemical Identifiers

CAS 7664-93-9, 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Colorless
Appearance of Solution Colorless Liquid
Concentration 17.95 to 18.05 N
CAS Min % 51.0, 47.0
CAS Max % 53.0, 48.92
Packaging Safety-coated Glass Bottle
Quantity 2.5 L
Physical Form Liquid
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
Grade Laboratory
Assay Normal at 20°C
Chemical Name or Material Sulfuric Acid
Show More Show Less
ShelfLife 36 months
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.