Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sulfuric Acid, 20% (v/v), Reagent Grade, Ricca Chemical

Shop All Ricca Chemical Company Products


Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Specifications
Specifications
Appearance of Solution | Colorless Liquid |
Concentration | 20% (v/v) |
CAS Min % | 79.0, 19.0 |
CAS Max % | 81.0, 21.0 |
Packaging | Natural Poly Bottle |
Solvent or Matrix | Aqueous Solution |
Quantity | 4 L |
Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
Percent Purity | 19.97 to 20.03% |
Grade | Reagent |
Show More |
Safety and Handling
ShelfLife | 36 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction