Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sulfuric Acid, 20% (w/w), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
20 L
Packaging:
Cubitainer
Safety-coated Amber Glass Bottle
Safety-coated Glass Bottle
Specifications
Specifications
Color | Colorless |
Concentration | 20% (w/w) |
CAS Min % | 79.0, 19.0 |
CAS Max % | 81.0, 21.0 |
Packaging | Safety-coated Amber Glass Bottle |
Solvent or Matrix | Aqueous Solution |
Quantity | 500 mL |
Physical Form | Liquid |
Percent Purity | 20% W/W |
Grade | Laboratory |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction