Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 50% v/v (1:1), AquaPhoenix Scientific™
SCI_ED

Catalog No. S88732 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle,Orange Cap
Poly Bottle,Purple Cap
Poly Bottle,White Cap
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S88732 500 mL Poly Bottle,Orange Cap
S88739 500 mL Poly Bottle,Purple Cap
S88738 500 mL Poly Bottle,White Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S88732 Supplier Aqua Phoenix Scientific Supplier No. SA1940P
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Concentration 50% v/v
Packaging Poly Bottle,Orange Cap
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.