Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sulfuric Acid, 50% (w/w), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
2.5 L
Specifications
Specifications
Packaging | Safety-coated Glass Bottle |
Quantity | 500 mL |
Percent Purity | 50% W/W |
Grade | Laboratory |
Chemical Name or Material | Sulfuric acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction