Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, ACS, 95.0-98.0%, Spectrum™ Chemical
SDP

Catalog No. 18614289 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
500 mL
2.5 L
5 gal.
185 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly-Coated Glass Bottle

Chemical Identifiers

CAS 7664-93-9

Specifications

CAS Max % 1
Heavy Metals 1 ppm
Packaging Drum
Residue after Ignition 5 ppm
Quantity 185 L
Percent Purity 95 to 98%
Grade ACS
WARNING: Cancer - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.