Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, ACS, 95.0-98.0%, Spectrum™ Chemical
SDP

Catalog No. 18614292 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
5 gal.
185 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly-Coated Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-292 5 gal. Drum
18-614-293 500 mL Amber Glass Bottle
18-614-290 2.5 L Poly-Coated Glass Bottle
18-614-289 185 L Drum
18-614-291 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-614-292 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S17305GLBL

Chemical Identifiers

CAS 7664-93-9

Specifications

CAS Max % 1
Heavy Metals 1 ppm
Packaging Drum
Residue after Ignition 5 ppm
Quantity 5 gal.
Percent Purity 95 to 98%
Grade Reagent
Chemical Name or Material Sulfuric Acid
WARNING: Cancer - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.