Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RABS003020E Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
20 L
Packaging:
Amber Glass Bottle
Hard Pack
Natural Poly Bottle
Safety-coated Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RABS003020E 20 L Hard Pack
RABS0030500 500 mL Amber Glass Bottle
RABS00301C 1 L Amber Glass Bottle
RABS003025D 2.5 L Safety-coated Glass Bottle
RABS00304C 4 L Amber Glass Bottle
RABS00304A 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. RABS003020E Supplier Ricca Chemical Company Supplier No. RABS003020E1
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Colorless
CAS Min % 95.0
CAS Max % 98.0
Heavy Metals (as Pb) max. 0.001%
Packaging Hard Pack
Reducing Substances Pass
Quantity 20 L
Synonym oil of vitriol
Percent Purity ≥85%
Grade ACS
Chemical Name or Material Sulfuric Acid
Show More Show Less
ShelfLife 36 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.