Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sulfuric Acid, ACS Reagent, Reagents
Click to view available options
Quantity:
1 L
2.5 L
75 lb.
750 lb.
Packaging:
Glass Bottle
HDPE Winpak
Poly Drum
Safety Coated Glass
Specifications
Specifications
pH | 0 |
Color | Colorless |
Boiling Point | 330°C |
Packaging | HDPE Winpak |
Specific Gravity | 1.84 |
Quantity | 75 lb. |
Physical Form | Liquid |
Grade | ACS |
Chemical Name or Material | Sulfuric Acid, ACS Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction