Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sulfuric acid concentrate, For 1 L standard solution, 0.5 M H2SO4 (1.0 N), Honeywell Fluka™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 Ea.
6 x 1 Ea.
Packaging:
Ampoule
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Boiling Point | >100°C |
| CAS Min % | ≥50.0000%, ≥25.0000% |
| CAS Max % | <75.0000%, <50.0000% |
| Density | 1.180 g/cm3 |
| Packaging | Ampoule |
| Quantity | 6 x 1 Ea. |
| Physical Form | Concentrate |
| Linear Formula | H2SO4 |
| UN Number | UN2796 |
| Beilstein | 2037554 |
| Show More |
Safety and Handling
| ShelfLife | 1260 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Sulfuric acid |
| Recommended Storage | Room Temp |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction