Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, For the determination of nitrogen, 97.5%, MilliporeSigma™ Supelco™

Catalog No. 111018399 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
11-101-8399 2.5 L Poly Bottle
11-101-9607 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8399 Supplier MilliporeSigma Supelco Supplier No. 847272.5L
Add to Cart
Add to Cart
Sulfuric acid can be produced by dry distillation of iron sulfate heptahydrate, copper sulfate pentahydrate. When these compounds were heated they decompose to form iron and copper oxides eliminating water and sulphur trioxide which combine together to form dilute solution of sulphuric acid. It is suitable as dehydrating agent, catalyst, active reactant in chemical industry. It is also suitable to control pH of saline solutions to strong fuming acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Clear
Boiling Point ˜290°C
Density 1.840 g/mL (at 25°C)
Packaging Poly Bottle
Vapor Pressure 1 mmHg (146°C)
Quantity 2.5 L
Linear Formula H2SO4
Formula Weight 98.08
Percent Purity ≥97.5%
RTECSNumber WS5600000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.