Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, NF, 95-98%, Spectrum™ Chemical
SDP

Catalog No. 18607172 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
2.5 L
6 x 500 mL
20 L
4 x 2.5 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Drum
Poly-Coated Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-172 4 x 2.5 L Poly-Coated Glass Bottle
18-607-171 2.5 L Glass Bottle
18-607-170 2.5 L Poly-Coated Glass Bottle
18-607-169 6 x 500 mL Amber Glass Bottle
18-607-173 4 x 2.5 L Glass Bottle
18-607-175 20 L Poly Drum
18-607-174 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-607-172 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S17312.5LTPVCS4
Add to Cart
Add to Cart

S1731, 7664-93-9, H2SO4

Spectrum™ Chemical Sulfuric Acid, NF is used as an acidifying agent in pharmaceutical applications. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

CAS Max % 1
Packaging Poly-Coated Glass Bottle
Quantity 4 x 2.5 L
Percent Purity 95 to 98%
Grade NF
Chemical Name or Material Sulfuric Acid
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.