Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid/Phosphoric Acid Mixture, Mill Titration Solution, Reagents

Catalog No. 01333702
Change view
Click to view available options
:
Quantity: 20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-333-702 Quantity: 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-702 Supplier Reagents Holdings Llc Supplier No. CS13480020E7
Only null left
Add to Cart
Add to Cart

Sulfuric Acid/Phosphoric Acid Mixture, Mill Titration Solution, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sulfuric Acid/Phosphoric Acid Mixture, Mill Titration Solution
pH 0
CAS 7732-18-5, 7664-93-9, 7664-38-2
Color Colorless
Quantity 20 L
Packaging HDPE Winpak
Physical Form Liquid
Specific Gravity 1.255
Molecular Formula H2SO4/H3PO4
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.