Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuryl chloride, 1.0M solution in dichloromethane, AcroSeal™

Catalog No. AC378158000
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle

CAS: 7791-25-5 | Cl2O2S | 134.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfuryl chloride
Name Note 1.0M Solution in Dichloromethane
CAS 7791-25-5, 75-09-2
Quantity 800 mL
Molecular Formula Cl2O2S
Linear Formula SO2Cl2
MDL Number MFCD00011451
Merck Index 15, 9110
Synonym sulfuryl chloride, sulfonyl chloride, sulphuryl dichloride, sulphuryl chloride, sulfonyl dichloride, sulfuric dichloride, sulfuric oxychloride, sulfurylchlorid, caswell no. 816, sulfurylchloride
InChI Key YBBRCQOCSYXUOC-UHFFFAOYSA-N
SMILES ClS(Cl)(=O)=O
IUPAC Name sulfuryl dichloride
Molecular Weight (g/mol) 134.96
PubChem CID 24648
ChEBI CHEBI:29291
Formula Weight 134.97
Concentration or Composition (by Analyte or Components) 1M, exact strength on the certificate of analysis
Packaging AcroSeal™ Glass Bottle
Appearance Clear pale yellow solution
Specific Gravity 1.352
Density 1.3520g/mL
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.