Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuryl chloride, 97%

Catalog No. AA89411A1
Click to view available options
Quantity:
250 g
1 kg

CAS: 7791-25-5 | Cl2O2S | 134.96 g/mol

Sulfuryl chloride is used as a chlorinating, sulfonating and chlorosulfonating agent. It is also used as a source of chlorine and acts as a reagent in the conversion of C-hydrogen to C-chlorine adjacent to activating substituent namely carbonyls and sulfoxides. As a chlorinating agent, it is involved in the chlorination of substrates like alkanes, alkenes, alkynes, aromatics, tetrahydrofuran and epoxides. It finds application in the conversion of thiols or disulfides into their corresponding sulfenyl chlorides. It plays a vital role to treat wool to prevent shrinkage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-25-5
Molecular Formula Cl2O2S
Molecular Weight (g/mol) 134.96
MDL Number MFCD00011451
InChI Key YBBRCQOCSYXUOC-UHFFFAOYSA-N
Synonym sulfuryl chloride, sulfonyl chloride, sulphuryl dichloride, sulphuryl chloride, sulfonyl dichloride, sulfuric dichloride, sulfuric oxychloride, sulfurylchlorid, caswell no. 816, sulfurylchloride
PubChem CID 24648
ChEBI CHEBI:29291
IUPAC Name sulfuryl dichloride
SMILES ClS(Cl)(=O)=O

Specifications

Boiling Point 69°C
Melting Point °C to 54°C
Physical Form Liquid
Quantity 1 kg
Assay Percent Range 97%
Solubility Information Miscible with benzene,toluene,chloroform,ether,carbon tetrachloride and glacial acetic acid.
Formula Weight 134.96
Refractive Index 1.4437
Sensitivity Moisture sensitive
Density 1.68
Chemical Name or Material Sulfuryl chloride
Show More Show Less
Hazard Category H314-H330-H335
Precautionary Statement P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: I
EINECSNumber 232-245-6
RTECSNumber WT4870000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.