Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfur pieces, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals

Catalog Number AA1034314
Click to view available options
Quantity:
25 g
100 g
500 g
7704-34-9
S
32.06
MFCD00085316
NINIDFKCEFEMDL-UHFFFAOYSA-N
polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot
5362487
CHEBI:26833
sulfur
[S]
7704-34-9
S
32.06
MFCD00085316
NINIDFKCEFEMDL-UHFFFAOYSA-N
polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot
5362487
CHEBI:26833
sulfur
[S]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Physical Form Pieces
Quantity 25 g
Assay Percent Range 99.9995% (Metals basis)
Ignition Point 248°C (478°F)
Odor Weak
Appearance Pale yellow pieces
Chemical Name or Material Sulphur
Hazard Category H315-H319-H500
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
DOTInformation DOT Class: 4.1, Packaging Group: III
EINECSNumber 231-722-6
RTECSNumber WS4250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit