Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SULT1A1 Monoclonal Antibody (OTI6C8), TrueMAB™, OriGene
SDP

Catalog No. 502541753
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1753 30 μL
50-167-3238 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1753 Supplier OriGene Supplier No. TA502159S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SULT1A1 Monoclonal antibody specifically detects SULT1A1 in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen SULT1A1
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI6C8
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene EXD1
Gene Accession No. Q8NHP7
Gene Alias AI266890; aryl sulfotransferase; Aryl sulfotransferase 1; Aryl sulfotransferase cytosolic 1A phenol-preferring member 3; Aryl sulfotransferase cytosolic, 1A, phenol-preferring, member 3; aryl sulfotransferase IV; arylsulfotransferase ST1A4; ASTIV; CSPS; HAST1/HAST2; MGC131921; MGC5163; Minoxidil sulfotransferase; mSTp1; Mx-ST; OK/SW-cl.88; OTTHUMP00000162568; OTTHUMP00000162569; P PST; P PST 1; Phenol sulfotransferase; Phenol sulfotransferase 1; Phenol sulfotransferase 1a1; phenol/aryl sulfotransferase; phenol-sulfating phenol sulfotransferase 1; P-PST; P-PST 1; PST; PST-1; St1a1; ST1A3; ST1A4; Stm; STP; STP1; sulfokinase; Sulfotransferase 1A1; sulfotransferase family 1A member 1; sulfotransferase family 1A, phenol-preferring, member 1; sulfotransferase family, cytosolic, 1A, phenol preferring; sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1; sulfotransferase, phenol preferring 1; sulphotransferase family cytosolic 1A phenol-prefering member 1; SULT1A1; Sult1a3; Thermostable phenol sulfotransferase; thermostable phenol sulfotransferase1; ts-PST; TSPST1; Tyrosine-ester sulfotransferase
Host Species Mouse
Immunogen Full length human recombinant protein of human EXD1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161829
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.