Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SULT1A1 Monoclonal Antibody (OTI9B7)
GREENER_CHOICE

Catalog No. PIMA525296
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525296 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525296 Supplier Invitrogen™ Supplier No. MA525296
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SULT1A1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

The soluble sulfotransferases contribute to the elimination of xenobiotics, the activation of procarcinogens and the regulation of hormones by catalyzing the sulfate conjugation of these substances. Members of the three groups comprising this superfamily (namely SULT1, SULT2 and SULT3) show selectivity to certain substrate compounds. SULT1A1 (sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1), also known as STP or STP1, is a 295 amino acid protein that localizes to the cytoplasm and belongs to the sulfotransferase family. Functioning as a homodimer that is expressed in brain, liver, skin and lung tissue, SULT1A1 catalyzes the sulfate conjugation of catecholamines, phenolic drugs and neurotransmitters and, via this catalytic activity, plays a role in the elimination of a variety of compounds from the body. Additionally, SULT1A1 may be involved in the activation of carcinogenic N-hydroxyarylamines, indicating a possible role in carcinogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SULT1A1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI9B7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SULT1A1
Gene Accession No. P50225
Gene Alias AI266890; aryl sulfotransferase; Aryl sulfotransferase 1; Aryl sulfotransferase cytosolic 1A phenol-preferring member 3; Aryl sulfotransferase cytosolic, 1A, phenol-preferring, member 3; aryl sulfotransferase IV; arylsulfotransferase ST1A4; ASTIV; CSPS; HAST1/HAST2; MGC131921; MGC5163; Minoxidil sulfotransferase; mSTp1; Mx-ST; OK/SW-cl.88; OTTHUMP00000162568; OTTHUMP00000162569; P PST; P PST 1; Phenol sulfotransferase; Phenol sulfotransferase 1; Phenol sulfotransferase 1a1; phenol/aryl sulfotransferase; phenol-sulfating phenol sulfotransferase 1; P-PST; P-PST 1; PST; PST-1; St1a1; ST1A3; ST1A4; Stm; STP; STP1; sulfokinase; Sulfotransferase 1A1; sulfotransferase family 1A member 1; sulfotransferase family 1A, phenol-preferring, member 1; sulfotransferase family, cytosolic, 1A, phenol preferring; sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1; sulfotransferase, phenol preferring 1; sulphotransferase family cytosolic 1A phenol-prefering member 1; SULT1A1; Sult1a3; Thermostable phenol sulfotransferase; thermostable phenol sulfotransferase1; ts-PST; TSPST1; Tyrosine-ester sulfotransferase
Gene Symbols SULT1A1
Host Species Mouse
Immunogen Full length human recombinant protein of SULT1A1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6817
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.