Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SULT1A1 Monoclonal Antibody (OTI9D10), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA501909

Description
Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. This gene encodes one of two phenol sulfotransferases with thermostable enzyme activity. Multiple alternatively spliced variants that encode two isoforms have been identified for this gene.Specifications
SULT1A1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P50225 | |
SULT1A1 | |
Full length human recombit protein of human SULT1A1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI9D10 | |
Unconjugated | |
SULT1A1 | |
HAST1/HAST2, P-PST, PST, ST1A1, ST1A3, STP, STP1, TSPST1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
6817 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction