Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SULT1A1 Mouse anti-Human, Clone: 10H6F6, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | SULT1A1 |
---|---|
Clone | 10H6F6 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6227
![]() ![]() |
Proteintech Group Inc
600761IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The soluble sulfotransferases contribute to the elimination of xenobiotics, the activation of procarcinogens and the regulation of hormones by catalyzing the sulfate conjugation of these substances. Members of the three groups comprising this superfamily (namely SULT1, SULT2 and SULT3) show selectivity to certain substrate compounds. SULT1A1 (sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1), also known as STP or STP1, is a 295 amino acid protein that localizes to the cytoplasm and belongs to the sulfotransferase family. Functioning as a homodimer that is expressed in brain, liver, skin and lung tissue, SULT1A1 catalyzes the sulfate conjugation of catecholamines, phenolic drugs and neurotransmitters and, via this catalytic activity, plays a role in the elimination of a variety of compounds from the body. Additionally, SULT1A1 may be involved in the activation of carcinogenic N-hydroxyarylamines, indicating a possible role in carcinogenesis.Specifications
SULT1A1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Aryl sulfotransferase 1, HAST1/HAST2, P PST, P PST 1, Phenol sulfotransferase 1, PST, ST1A1, ST1A3, STP, STP1, Sulfotransferase 1A1, SULT1A1, Ts PST, TSPST1 | |
SULT1A1 | |
IgG2a | |
Caprylic Acid Precipitation | |
Antibody |
10H6F6 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P50225 | |
6817 | |
SULT1A1 Fusion Protein Ag4611 | |
Primary | |
-20°C | |
SULT1A1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title