Learn More
SULT1A1 Mouse anti-Human, Clone: OTI2C5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501952

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. This gene encodes one of two phenol sulfotransferases with thermostable enzyme activity. Multiple alternatively spliced variants that encode two isoforms have been identified for this gene.Specifications
SULT1A1 | |
Monoclonal | |
Unconjugated | |
SULT1A1 | |
HAST1/HAST2, P-PST, PST, ST1A1, ST1A3, STP, STP1, TSPST1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
6817 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI2C5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P50225 | |
SULT1A1 | |
Full length human recombit protein of human SULT1A1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.